Anti-SLC36A2 Rabbit Polyclonal Antibody
Catalog # 77120-954
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:solute carrier family 36 (proton/amino acid symporter), member 2
- Antigen Symbol:SLC36A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:153201
- Antigen Synonyms:proton-coupled amino acid transporter 2|PAT2|Solute carrier family 36 (proton/amino acid symporter) member 2|TRAMD1
- Modification:None
- Storage Buffer:SLC36A2 antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:SLC36A2 antibody can be stored at 4...730 °C for three months and –20...+730 °C, stable for up to one year.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:SLC36A2 antibody was raised against a 15 amino acid peptide near the amino terminus of human SLC36A2.The immunogen is located within amino acids 30-80 of SLC36A2.
- Purification:SLC36A2 antibody is affinity chromatography purified via peptide column.
- Cat. No.:77120-954
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SLC36A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human