Anti-TRPC3 Rabbit Polyclonal Antibody
Catalog # 77126-972
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:transient receptor potential cation channel, subfamily C, member 3
- Antigen Symbol:TRPC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Form:Liquid
- Gene ID:7222
- Antigen Synonyms:Transient receptor protein 3|TrpC3|TRP3|Short transient receptor potential channel 3
- Modification:None
- Storage Buffer:TRPC3 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:TRPC3 antibody can be stored at 4...730 °C for three months and –20...+730 °C, stable for up to one year.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:TRPC3 antibody was raised against a 19 amino acid peptide from near the carboxy terminus of human TRPC3.The immunogen is located within amino acids 710-760 of TRPC3.
- Purification:TRPC3 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:77126-972
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TRPC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat