Anti-AJAP1 Rabbit Polyclonal Antibody
Catalog # 77119-302
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:adherens junctions associated protein 1
- Antigen Symbol:AJAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:55966
- Antigen Synonyms:SHREW1|Membrane protein shrew-1|MOT8|Adherens junctions associated protein 1
- Modification:None
- Storage Buffer:AJAP1 antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:AJAP1 antibody can be stored at 4...730 °C for three months and –20...+730 °C, stable for up to one year.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:AJAP1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human AJAP1.The immunogen is located within amino acids 290-340 of AJAP1.
- Purification:AJAP1 antibody is affinity chromatography purified via peptide column.
- Cat. No.:77119-302
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: AJAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat