Order Entry
United States
Orders LinkContactUsLinkComponent
 

GNIP Peptide

Vial, 0.1 mg
Catalog # 77125-654
Supplier:   Prosci
 :  45-684P
GNIP Peptide
Catalog # 77125-654
Supplier:  Prosci
CAS Number:  
GNIP Peptide

Vial, 0.1 mg

CAS Number:  
Price
$301.99
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    0.1 mg
  • Storage Conditions:
    Store at −20 °C. When reconstituted minimize freezing and thawing.
  • Cat. No.:
    77125-654
  • Shipping Temperature:
    RT
  • Protein/Peptide Type:
    Synthetic
  • Gene ID:
    81786
  • Formulation:
    Lyophilized. GNIP peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
  • Tested Applications:
    Blocking
  • Protein Synonyms:
    RNF90|GNIP|Glycogenin-interacting protein|Tripartite motif-containing protein 7
  • Protein/Peptide Name:
    TRIM7
  • Conjugation:
    Unconjugated
  • Species:
    Human

Specifications