Cortactin Peptide
77121-768
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Mouse
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:2017
- Protein Synonyms:EMS1|Src substrate cortactin|Amplaxin
- Protein/Peptide Name:CTTN
- Formulation:Lyophilized. Cortactin peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77121-768