Order Entry
United States
ContactUsLinkComponent
Human Recombinant HGF Protein (from HEK293 cells)
Human Recombinant HGF Protein (from HEK293 cells)
Catalog # 77130-390
Supplier:  Prosci
CAS Number:  
Human Recombinant HGF Protein (from HEK293 cells)
Catalog # 77130-390
Supplier:  Prosci
Supplier Number:  40-627
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    5 µg
  • Storage Conditions:
    The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
  • Biological Activity:
    Determined by the dose-dependent stimulation of the proliferation of monkey 4MBr-5 cells.
  • Gene ID:
    3082
  • Protein Synonyms:
    Scatter Factor (SF)|Hepatopoietin (HPTA)|Hepatocyte Growth Factor
  • Protein/Peptide Name:
    HGF
  • Purity:
    ≥95% by SDS-PAGE gel and HPLC analyses
  • Sequence:
    alpha chain: KRRNTIHEFK KSAKTTLIKI DPALKIKTKK VNTADQCANR CTRNKGLPFT CKAFVFDKAR KQCLWFPFNS MSSGVKKEFG HEFDLYENKD YIRNCIIGKG RSYKGTVSIT KSGIKCQPWS SMIPHEHSFL PSSYRGKDLQ ENYCRNPRGE EGGPWCFTSN PEVRYEVCDI PQCSEVECMT CNGESYRGLM DHTESGKICQ RWDHQTPHRH KFLPERYPDK GFDDNYCRNP DGQPRPWCYT LDPHTRWEYC AIKTCADNTM NDTDVPLETT ECIQGQGEGY RGTVNTIWNG IPCQRWDSQY PHEHDMTPEN FKCKDLRENY CRNPDGSESP WCFTTDPNIR VGYCSQIPNC DMSHGQDCYR GNGKNYMGNL SQTRSGLTCS MWDKNMEDLH RHIFWEPDAS KLNENYCRNP DDDAHGPWCY TGNPLIPWDY CPISRCEGDT TPTIVNLDHP VISCAKTKQL R

    beta chain: VVNGIPTRTN IGWMVSLRYR NKHICGGSLI KESWVLTARQ CFPSRDLKDY EAWLGIHDVH GRGDEKCKQV LNVSQLVYGP EGSDLVLMKL ARPAVLDDFV STIDLPNYGC TIPEKTSCSV YGWGYTGLIN YDGLLRVAHL YIMGNEKCSQ HHRGKVTLNE SEICAGAEKI GSGPCEGDYG GPLVCEQHKM RMVLGVIVPG RGCAIPNRPG IFVRVAYYAK WIHKIILTYK VPQS
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    RT
  • Cat. No.:
    77130-390

Specifications

About this item