SARS-CoV-2 (COVID-19, 2019-nCoV) Recombinant Full-Length Epsilon Variant (B.1.429, Ca) Spike Protein (from HEK293 cells)
Catalog # 77124-944
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Size:0.025 mg
- Storage Conditions:–80 °C. Avoid freeze-thaw cycles.
- Biological Activity:Not determined
- Gene ID:43740568
- Protein Synonyms:SARS-CoV-2 (COVID-19) Spike Recombinant Protein: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)|Spike protein|Surface Glycoprotein
- Protein/Peptide Name:Spike
- Purity:>98% as determined by SDS-PAGE. PureCube Rho1D4 Agarose.
- Molecular Weight:1286 amino acids (including Rho1D4 tag and linker); 142115.37 Da
- Sequence:Full-length sequence (aa 1 – 1273): S13I; W152C; L452R; D1183Y furin cleavage site “RRAR” mutated to “GSAG”; K986P; V987P C-terminal Rho1D4 tag fused with spacer “GSSG” to protein sequence
- Concentration:Batch dependent
- Formulation:Liquid. 20 mM Hepes pH 7.5; 150 mM NaCl, 0.001% LMNG
- Shipping Temperature:Dry Ice
- Tested Applications:ELISA
- Cat. No.:77124-944