Human Recombinant SARS-CoV-2 (COVID-19) Gamma Variant (P.1, Brazil) Spike S1 (RBD) Variant Protein (from HEK293 cells)
Catalog # 77130-802
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:Short term storageL: +4 °C, Long term storage: −20 °C, Handling Advice: After opening, prepare aliquots and store at −20 °C. Avoid freeze/thaw cycles. Centrifuge lyophilized vial before opening and reconstitution. For maximum product recovery after thawing, centrifuge the vial before opening the cap. Use/Stability: Stable for at least 6 months after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Biological Activity:Binds to anti-SARS-CoV-2 Spike (RBD) antibodies in serum or plasma. Binds to human ACE2.
- Gene ID:43740568
- Protein Synonyms:2019-nCoV Spike Protein S1 (RBD)|Spike Receptor Binding Domain|VOC-202101/02|20J/501Y.V3
- Protein/Peptide Name:Spike
- Purity:≥95% (SDS-PAGE)
- Molecular Weight:~35 kDa (SDS-PAGE)
- Sequence:Receptor-binding domain (RBD) of SARS-CoV-2 Spike protein S1 (aa 319-541) containing the mutations K417T, E484K, N501Y is fused at the C-terminus to a His-tag.
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilized. Lyophilized. Contains PBS.
- Shipping Temperature:RT
- Cat. No.:77130-802