SARS Recombinant Coronavirus Envelope (HSZ-Cc) Protein (from bacteria)
Catalog # 77134-020
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Bacteria
- Species:SARS
- Size:0.1 mg
- Storage Conditions:Store at −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:1489671
- Protein Synonyms:E Protein SARS-CoV HSZ-Cc|Envelope Protein SARS-CoV HSZ-Cc|SARS-CoV HSZ-Cc Envelope Protein|SARS-CoV HSZ-Cc-E Protein
- Protein/Peptide Name:Envelope
- Purity:GSH-Agarose (0.65)
- Molecular Weight:Mono-Isotopic Mass: 35, 162.15 daltons Average Mass: 35, 185.17 daltons
- Sequence:MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDLEVLFQGPLGSMYSFVSEETGTLIVNSVLLFLAFVVFLLVTLAILTALRLCAYCCNIVNVSLVKPTVYVYSRVKNLNSSEGVPDLLV Amino acids M1 – V76 (end). Residue M232 of the fusion protein is equivalent to M1 of the native enzyme. The GST tag is located at residues 1 – 220. Protease Cleavage: PreScission (LEVLFQGP) residues 221 - 228
- Concentration:Batch dependent
- Formulation:Liquid. 50 mM Tris-HCl pH 7.5, 270 mM Sucrose, 150 mM NaCl, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35
- Shipping Temperature:Dry Ice
- Cat. No.:77134-020