- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Bacteria
- Species:SARS
- Size:0.1 mg
- Environmentally Preferable:
- Storage Conditions:Store at −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:1489678
- Protein Synonyms:Nucleocapsid Protein SARS CoV HSZ-Cc|N Protein SARS CoV HSZ-Cc|SARS CoV HSZ-Cc Nucleocapsid Protein|SARS-CoV-HSZ-Cc-N Protein
- Protein/Peptide Name:Nucleocapsid
- Purity:Amylose Resin (0.85)
- Molecular Weight:Mono-Isotopic Mass: 90, 208.50 daltons Average Mass: 90, 263.94 daltons
- Concentration:Batch dependent
- Formulation:Liquid. 50 mM Tris-HCl pH 7.5, 270 mM Sucrose, 150 mM NaCl, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35
- Shipping Temperature:Dry Ice
- Cat. No.:77134-530
Native Sequence: MKIEEGKLVIWINGDKGYNGLAEVGKKFEKDTGIKVTVEHPDKLEEKFPQVAATGDGPDIIFWAHDRFGGYAQSGLLAEITPDKAFQDKLYPFTWDAVRYNGKLIAYPIAVEALSLIYNKDLLPNPPKTWEEIPALDKELKAKGKSALMFNLQEPYFTWPLIAADGGYAFKYENGKYDIKDVGVDNAGAKAGLTFLVDLIKNKHMNADTDYSIAEAAFNKGETAMTINGPWAWSNIDTSKVNYGVTVLPTFKGQPSKPFVGVLSAGINAASPNKELAKEFLENYLLTDEGLEAVNKDKPLGAVALKSYEEELVKDPRIAATMENAQKGEIMPNIPQMSAFWYAVRTAVINAASGRQTVDEALKDAQTNSSSNNNNNNNNNNLGDDDDKVPEFLEVLFQGPLGSMSDNGPQSNQRSAPRITFGGPTDSTDNNQNGGRNGARPKQRRPQGLPNNTASWFTALTQHGKEELRFPRGQGVPINTNSGPDDQIGYYRRATRRVRGGDGKMKELSPRWYFYYLGTGPEASLPYGANKEGIVWVATEGALNTPKDHIGTRNPNNNAATVLQLPQGTTLPKGFYAEGSRGGSQASSRSSSRSRGNSRNSTPGSSRGNSPARMASGGGETALALLLLDRLNQLESKVSGKGQQQQGQTVTKKSAAEASKKPRQKRTATKQYNVTQAFGRRGPEQTQGNFGDQDLIRQGTDYKHWPQIAQFAPSASAFFGMSRIGMEVTPSGTWLTYHGAIKLDDKDPQFKDNVILLNKHIDAYKTFPPTEPKKDKKKKTDEAQPLPQRQKKQPTVTLLPAADMDDFSRQLQNSMSGASADSTQA Residue M404 of the fusion protein is equivalent to M1 of the native enzyme. The MBP tag is located at residues 1 – 367. Protease Cleavage: PreScission (LEVLFQGP) residues 393 – 400