Human Recombinant CD96/TACTILE Protein (from HEK293 cells)
Catalog # 77128-968
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:10225
- Protein Synonyms:TACTILE
- Protein/Peptide Name:CD96
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:55.4 kDa
- Sequence:Val 22 - Met 503
- Formulation:Lyophilized. PBS, pH 7.4
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77128-968