Anti-CUX1 Rabbit Polyclonal Antibody
Catalog # 76749-578
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Cut-like homeobox 1
- Antigen Symbol:CUX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:1523
- Antigen Synonyms:CUX1|CUTL1|CUT|p75|Cut-like homeobox 1|CASP|p200|CUX|p100|GOLIM6|Cut homolog|p110|Nbla10317|Homeobox protein cux-1|COY1|CDP|Clox|CDP1|Protein CASP|CCAAT displacement protein
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CUX1 (NP_001189472.1).
- Purification:Affinity purification
- Cat. No.:76749-578
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CUX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat