Anti-KBTBD3 Rabbit Polyclonal Antibody
Catalog # 76751-672
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Kelch repeat and BTB (POZ) domain containing 3
- Antigen Symbol:KBTBD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:69149
- Antigen Synonyms:BTB and kelch domain containing 3|BTB and kelch domain-containing protein 3|kelch repeat and BTB domain-containing protein 3|2200003A07Rik|Bklhd3
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 410-607 of mouse KBTBD3 (NP_081238.2).
- Purification:Affinity purification
- Cat. No.:76751-672
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: KBTBD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human