Anti-APBB1 Rabbit Polyclonal Antibody
Catalog # 76754-712
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65)
- Antigen Symbol:APBB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:322
- Antigen Synonyms:amyloid beta A4 precursor protein-binding|APBB1|FE65|adaptor protein FE65a2|stat-like protein|MGC:9072|member 1|amyloid beta (A4) precursor protein-binding|RIR|family B|member 1 (Fe65)
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 359-708 of human APBB1 (NP_663722.1).
- Purification:Affinity purification
- Cat. No.:76754-712
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: APBB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human