Anti-GLB1 Rabbit Polyclonal Antibody
Catalog # 76756-540
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Beta-galactosidase
- Antigen Symbol:GLB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:2720
- Antigen Synonyms:Acid beta-galactosidase|Lactase|Elastin receptor 1|Beta-galactosidase|GLB1|ELNR1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human GLB1 (NP_001073279.1).
- Purification:Affinity purification
- Cat. No.:76756-540
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: GLB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat