Anti-HILPDA Rabbit Polyclonal Antibody
Catalog # 76751-724
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Hypoxia inducible lipid droplet associated
- Antigen Symbol:HILPDA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:29923
- Antigen Synonyms:hypoxia inducible gene 2|C7orf68|HIG-2|hypoxia-inducible protein 2|hypoxia-inducible lipid droplet-associated protein|hypoxia-inducible gene 2 protein|HIG2
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant protein of human HILPDA.
- Purification:Affinity purification
- Cat. No.:76751-724
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HILPDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human