Anti-BMPR2 Rabbit Polyclonal Antibody
Catalog # 76757-310
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Bone morphogenetic protein receptor type II
- Antigen Symbol:BMPR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:659
- Antigen Synonyms:BMPR-II|type II (serine/threonine kinase)|PPH1|TRG10|transformatio|BRK-3|FLJ76945|bone morphogenetic protein receptor type II|BMR2|serine/threonine kinase|T-ALK|bone morphogenetic protein receptor|BMPR3|primary pulmonary hypertension 1|BMPR2
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 27-150 of human BMPR2 (NP_001195.2).
- Purification:Affinity purification
- Cat. No.:76757-310
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human