Anti-HSP90AB1 Rabbit Polyclonal Antibody
Catalog # 76749-530
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Heat shock protein 90 kDa alpha (cytosolic), class B member 1
- Antigen Symbol:HSP90AB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3326
- Antigen Synonyms:HSP84|D6S182|HSPCB|HSPC2|HSP90B|HSP90AB1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human HSP90AB1 (NP_031381.2).
- Purification:Affinity purification
- Cat. No.:76749-530
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey