Anti-GPR153 Rabbit Polyclonal Antibody
Catalog # 76762-782
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:G protein-coupled receptor 153
- Antigen Symbol:GPR153
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:387509
- Antigen Synonyms:PGR1|G protein-coupled receptor 153|GPR153
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide of Human GPR153.
- Purification:Affinity purification
- Cat. No.:76762-782
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: GPR153
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse