Anti-VWC2 Rabbit Polyclonal Antibody
Catalog # 76751-268
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Von Willebrand factor C domain containing 2
- Antigen Symbol:VWC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Other,Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:319922
- Antigen Synonyms:UNQ739|cradin|G11|von Willebrand factor C domain-containing protein 2|brorin|CR (chordin-like cysteine-rich) domain-containing adhesive protein|PSST739|A930041G11Rik|brain-specific chordin-like protein
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein of Human VWC2.
- Purification:Affinity purification
- Cat. No.:76751-268
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: VWC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Other