Anti-DNAJC18 Rabbit Polyclonal Antibody
Catalog # 76750-942
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:DnaJ heat shock protein family (Hsp40) member C18
- Antigen Symbol:DNAJC18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:202052
- Antigen Synonyms:member 18|DnaJ (Hsp40) homolog|subfamily C|dnaJ homolog subfamily C member 18
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 260-350 of human DNAJC18 (NP_689899.1).
- Purification:Affinity purification
- Cat. No.:76750-942
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: DNAJC18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat