Anti-IGSF11 Rabbit Polyclonal Antibody
Catalog # 76761-552
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Immunoglobulin superfamily, member 11
- Antigen Symbol:IGSF11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Other,Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:152404
- Antigen Synonyms:IGSF11|BT-IgSF|MGC35227|CXADRL1|Igsf13|CT119|VSIG3
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 80-200 of human IGSF11 (NP_689751.2).
- Purification:Affinity purification
- Cat. No.:76761-552
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: IGSF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Other