Anti-N4BP2L1 Rabbit Polyclonal Antibody
76753-098
: Prosci
- Antibody Type:Primary
- Antigen Name:NEDD4 binding protein 2 like 1
- Antigen Symbol:N4BP2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:90634
- Antigen Synonyms:NEDD4-binding protein 2-like 1|CG018|hypothetical gene CG018
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human N4BP2L1 (NP_001073159).
- Purification:Affinity purification
- Cat. No.:76753-098
- Packaging:Vial
Type: Primary
Antigen: N4BP2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse