Anti-SCIN Rabbit Polyclonal Antibody
Catalog # 76764-672
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Scinderin
- Antigen Symbol:SCIN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:85477
- Antigen Synonyms:adseverin
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide of Human SCIN.
- Purification:Affinity purification
- Cat. No.:76764-672
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SCIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human