Anti-PRKAA1/PRKAA2 Rabbit Polyclonal Antibody
Catalog # 76754-990
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen Symbol:PRKAA1/PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:5562;5563
- Antigen Synonyms:AMPK|HMG-CoA redustase kinase|VEGFR2|AMPK-alpha1|PRKAA1|AAPK1|AMPKa1|AMPK alpha-1 chain|FLK1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant protein of human PRKAA1/PRKAA2.
- Purification:Affinity purification
- Cat. No.:76754-990
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PRKAA1/PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat