Anti-P2RY12 Rabbit Polyclonal Antibody
Catalog # 76756-608
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Purinergic receptor P2Y12
- Antigen Symbol:P2RY12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:64805
- Antigen Synonyms:P2Y(12)R|ADPG-R|ADP-glucose receptor|BDPLT8|G-protein coupled|P2T(AC)|P2Y12|P2Y purinoceptor 12|SP1999|Gi-coupled ADP receptor HORK3|P2Y(AC)|P2Y12 platelet ADP receptor|P2Y(ADP)|12|purinergic receptor P2Y|purinergic receptor P2RY12|putative G-protein coupled receptor|HORK3|P2Y(cyc)|G-protein coupled receptor SP1999
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human P2RY12 (NP_795345.1).
- Purification:Affinity purification
- Cat. No.:76756-608
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: P2RY12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat