Anti-IFNA21 Rabbit Polyclonal Antibody
Catalog # 76750-872
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon alpha 21
- Antigen Symbol:IFNA21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3452
- Antigen Synonyms:leIF-F|interferon alpha-21|IFN-alphaI|IFN-alpha-21|leukocyte interferon protein|interferon alpha-F|LeIF F
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human IFNA21 (NP_002166.2).
- Purification:Affinity purification
- Cat. No.:76750-872
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: IFNA21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse