Anti-ZNF76 Rabbit Polyclonal Antibody
Catalog # 76756-432
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Zinc finger protein 76
- Antigen Symbol:ZNF76
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:7629
- Antigen Synonyms:Zfp523|D6S229E|ZNF76|ZNF523
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 100-170 of human ZNF76 (NP_003418.2).
- Purification:Affinity purification
- Cat. No.:76756-432
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ZNF76
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat