Anti-DHX57 Rabbit Polyclonal Antibody
Catalog # 76754-736
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:DExH-box helicase 57
- Antigen Symbol:DHX57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:90957
- Antigen Synonyms:DEAH box protein 57|DEAH-box RNA/DNA helicase AAM73547|DEAH-box helicase 57|DDX57|DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57|putative ATP-dependent RNA helicase DHX57
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 700-900 of human DHX57 (NP_945314.1).
- Purification:Affinity purification
- Cat. No.:76754-736
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: DHX57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat