Anti-KLF8 Rabbit Polyclonal Antibody
Catalog # 76761-254
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Kruppel-like factor 8
- Antigen Symbol:KLF8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:11279
- Antigen Synonyms:ZNF741|BKLF3|KLF8
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 210-280 of human KLF8 (NP_009181.2).
- Purification:Affinity purification
- Cat. No.:76761-254
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: KLF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat