Anti-CHPT1 Rabbit Polyclonal Antibody
Catalog # 76754-952
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Choline phosphotransferase 1
- Antigen Symbol:CHPT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:56994
- Antigen Synonyms:hCPT1|cholinephosphotransferase 1 alpha|AAPT1-like protein|CPT1|cholinephosphotransferase 1|CPT|diacylglycerol cholinephosphotransferase 1|phosphatidylcholine synthesizing enzyme
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CHPT1 (NP_064629.2).
- Purification:Affinity purification
- Cat. No.:76754-952
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CHPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human