Anti-PRSS21 Rabbit Polyclonal Antibody
Catalog # 76753-336
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Serine protease 21
- Antigen Symbol:PRSS21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:10942
- Antigen Synonyms:TESTISIN|ESP-1|serine protease from eosinophils|ESP1|protease|testisin|eosinophil serine protease 1|serine 21|TEST1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 150-220 of human PRSS21 (NP_006790.1).
- Purification:Affinity purification
- Cat. No.:76753-336
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PRSS21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat