Anti-KCNK6 Rabbit Polyclonal Antibody
76753-462
: Prosci
- Antibody Type:Primary
- Antigen Name:Potassium channel subfamily K member 6
- Antigen Symbol:KCNK6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:9424
- Antigen Synonyms:Potassium channel subfamily K member 6|TWIK-originated similarity sequence|TWIK2|Inward rectifying potassium channel protein TWIK-2|KCNK6|TOSS
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 254-313 of human KCNK6 (NP_004814.1).
- Purification:Affinity purification
- Cat. No.:76753-462
- Packaging:Vial
Type: Primary
Antigen: KCNK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat