Anti-ANT1/ANT2/ANT3/ANT4 Rabbit Polyclonal Antibody
Catalog # 76760-382
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Symbol:ANT1/ANT2/ANT3/ANT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:291;292;293;83447
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human ANT1 (NP_001142.2).
- Purification:Affinity purification
- Cat. No.:76760-382
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ANT1/ANT2/ANT3/ANT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat