Anti-TIGD1 Rabbit Polyclonal Antibody
Catalog # 76761-916
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Tigger transposable element derived 1
- Antigen Symbol:TIGD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:200765
- Antigen Synonyms:tigger transposable element derived 1|TIGD1|EEYORE|jerky (mouse) homolog-like
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TIGD1 (NP_663748.1).
- Purification:Affinity purification
- Cat. No.:76761-916
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TIGD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat