Anti-SSC4D Rabbit Polyclonal Antibody
Catalog # 76761-626
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Scavenger receptor cysteine-rich domain-containing group B protein {ECO:0000303|PubMed:12466895, ECO:0000312|HGNC:HGNC:14461}
- Antigen Symbol:SSC4D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:136853
- Antigen Synonyms:ECO:0000312|HGNC:HGNC:14461}|Scavenger receptor cysteine-rich domain-containing group B protein {ECO:0000303|PubMed:12466895|S4D-SRCRB|Four scavenger receptor cysteine-rich domains-containing protein {ECO:0000312|HGNC:HGNC:14461}|SSC4D
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 446-575 of human SSC4D (NP_542782.1).
- Purification:Affinity purification
- Cat. No.:76761-626
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SSC4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat