Anti-CHCHD1 Rabbit Polyclonal Antibody
Catalog # 76758-720
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Coiled-coil-helix-coiled-coil-helix domain containing 1
- Antigen Symbol:CHCHD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:118487
- Antigen Synonyms:mitochondrial small ribosomal subunit protein mS37|C236MRP-S37|mitochondrial|nuclear protein C2360|C10orf34|28S ribosomal protein S37|coiled-coil-helix-coiled-coil-helix domain-containing protein 1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human CHCHD1 (NP_976043.1).
- Purification:Affinity purification
- Cat. No.:76758-720
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CHCHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse