Anti-RAB11FIP4 Rabbit Polyclonal Antibody
76760-924
: Prosci
- Antibody Type:Primary
- Antigen Name:RAB11 family interacting protein 4
- Antigen Symbol:RAB11FIP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:84440
- Antigen Synonyms:RAB11-FIP4|arfophilin-2|RAB11 family interacting protein 4 (class II)|FIP4-Rab11|rab11 family-interacting protein 4
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 338-637 of human RAB11FIP4 (NP_116321.2).
- Purification:Affinity purification
- Cat. No.:76760-924
- Packaging:Vial
Type: Primary
Antigen: RAB11FIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human