Anti-EXOSC5 Rabbit Polyclonal Antibody
Catalog # 76756-216
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Exosome component 5
- Antigen Symbol:EXOSC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:56915
- Antigen Synonyms:p12B|exosome complex exonuclease RRP46|chronic myelogenous leukemia tumor antigen 28|RRP41B|Rrp46p|exosome component Rrp46|exosome complex component RRP46|hRrp46p|ribosomal RNA-processing protein 46|RRP46
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-235 of human EXOSC5 (NP_064543.3).
- Purification:Affinity purification
- Cat. No.:76756-216
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: EXOSC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat