Anti-SAFB Rabbit Polyclonal Antibody
Catalog # 76763-008
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Scaffold attachment factor B
- Antigen Symbol:SAFB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:6294
- Antigen Synonyms:SAFB|HET|SAB-B1|SAFB1|SAF-B1|Scaffold attachment factor B|SAF-B|Scaffold attachment factor B1|HSP27 ERE-TATA-binding protein|Hsp27 ERE-TATA binding protein|HAP
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 300-400 of human SAFB (NP_002958.2).
- Purification:Affinity purification
- Cat. No.:76763-008
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SAFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat