Anti-CDC20 Rabbit Polyclonal Antibody
76756-938
: Prosci
- Antibody Type:Primary
- Antigen Name:Cell division cycle protein 20 homolog
- Antigen Symbol:CDC20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:991
- Antigen Synonyms:Cell division cycle protein 20 homolog|p55CDC|CDC20
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CDC20 (NP_001246.2).
- Purification:Affinity purification
- Cat. No.:76756-938
- Packaging:Vial
Type: Primary
Antigen: CDC20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat