Anti-LSM11 Rabbit Polyclonal Antibody
76751-114
: Prosci
- Antibody Type:Primary
- Antigen Name:LSM11, U7 small nuclear RNA associated
- Antigen Symbol:LSM11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:134353
- Antigen Synonyms:U7 snRNA-associated Sm-like protein LSm11
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 131-360 of human LSM11 (NP_775762.1).
- Purification:Affinity purification
- Cat. No.:76751-114
- Packaging:Vial
Type: Primary
Antigen: LSM11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human