Anti-JDP2 Rabbit Polyclonal Antibody
76749-798
: Prosci
- Antibody Type:Primary
- Antigen Name:Jun dimerization protein 2
- Antigen Symbol:JDP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:122953
- Antigen Synonyms:jun dimerization protein 2|JUNDM2|progesterone receptor co-activator
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-174 of human JDP2 (NP_001128521.1).
- Purification:Affinity purification
- Cat. No.:76749-798
- Packaging:Vial
Type: Primary
Antigen: JDP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat