Anti-NDUFC2 Rabbit Polyclonal Antibody
76748-676
: Prosci
- Antibody Type:Primary
- Antigen Name:NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5 kDa
- Antigen Symbol:NDUFC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:4718
- Antigen Synonyms:B14.5b|CI-B14.5b|NADHDH2|HLC-1|NDUFC2
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NDUFC2 (NP_004540.1).
- Purification:Affinity purification
- Cat. No.:76748-676
- Packaging:Vial
Type: Primary
Antigen: NDUFC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat