Anti-GOLGA7 Rabbit Polyclonal Antibody
76755-126
: Prosci
- Antibody Type:Primary
- Antigen Name:Golgin A7
- Antigen Symbol:GOLGA7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:51125
- Antigen Synonyms:golgi complex-associated protein of 16 kDa|GOLGA7|GOLGA3AP1A|golgin subfamily a|7|GCP16|HSPC041|golgi autoantigen|golgin subfamily A member 7|Golgi complex-associated protein of 16kDa
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of human GOLGA7 (NP_001167595.1).
- Purification:Affinity purification
- Cat. No.:76755-126
- Packaging:Vial
Type: Primary
Antigen: GOLGA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse