Anti-RNASE12 Rabbit Polyclonal Antibody
76761-338
: Prosci
- Antibody Type:Primary
- Antigen Name:Ribonuclease A family member 12 (inactive)
- Antigen Symbol:RNASE12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:493901
- Antigen Synonyms:ribonuclease A I1|RAI1|epididymis secretory sperm binding protein Li 85p|HEL-S-85p|probable inactive ribonuclease-like protein 12|ribonuclease|RNase A family|ribonuclease-like protein 12|12 (non-active)
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RNASE12 (NP_001019993.1).
- Purification:Affinity purification
- Cat. No.:76761-338
- Packaging:Vial
Type: Primary
Antigen: RNASE12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human