Anti-CORO2A Rabbit Polyclonal Antibody
Catalog # 76752-364
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Coronin-2A
- Antigen Symbol:CORO2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:7464
- Antigen Synonyms:IR10|WD repeat-containing protein 2|Coronin-2A|WDR2|CORO2A
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 376-525 of human CORO2A (NP_003380.3).
- Purification:Affinity purification
- Cat. No.:76752-364
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CORO2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat