Anti-NEURL2 Rabbit Polyclonal Antibody
Catalog # 76753-298
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Neuralized E3 ubiquitin protein ligase 2
- Antigen Symbol:NEURL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:140825
- Antigen Synonyms:Ozz-E3|C20orf163|MGC125934|MGC125935|OZZ-E3|NEURL2|OZZ
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 181-285 of human NEURL2 (NP_542787.1).
- Purification:Affinity purification
- Cat. No.:76753-298
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: NEURL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat