Anti-ATP6V1G3 Rabbit Polyclonal Antibody
Catalog # 76763-316
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:ATPase H+ transporting V1 subunit G3
- Antigen Symbol:ATP6V1G3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:127124
- Antigen Synonyms:Vma1V-type proton ATPase subunit G 3|V1 subunit G3|vacuolar proton pump|V-ATPase G subunit 3|V-ATPase subunit G 3|ATP6G3|lysosomal 13kDa|vacuolar proton pump subunit G 3|vacuolar ATP synthase subunit G 3|lysosomal (vacuolar proton pump) subunit G3|V-ATPase 13 kDa subunit 3|vacuolar proton pump G subunit 3|H+ transporting|ATPase|subunit G3|V-ATPase G3 subunit
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human ATP6V1G3 (NP_573569.1).
- Purification:Affinity purification
- Cat. No.:76763-316
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ATP6V1G3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse