Anti-SLC22A23 Rabbit Polyclonal Antibody
Catalog # 76750-572
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Solute carrier family 22, member 23
- Antigen Symbol:SLC22A23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:63027
- Antigen Synonyms:FLJ22174|DKFZP434F011|C6orf85|SLC22A23
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 336-405 of human SLC22A23 (NP_068764.3).
- Purification:Affinity purification
- Cat. No.:76750-572
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SLC22A23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat